(2-Bromobenzylidene)malononitrile

Lieferant: Apollo Scientific

OR111250-1G
APOSOR111250-1GEA 0 EUR
APOSOR111250-1G
(2-Bromobenzylidene)malononitrile
(2-Bromobenzylidene)malononitrile
Formel: C₁₀H₅BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00175208
CAS-Nummer: 2698-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR