Methyl 4-(2,2-dicyanovinyl)benzoate
Lieferant: Apollo Scientific
|
Achtung
|
OR111251-5G
APOSOR111251-5GEA
0
EUR
APOSOR111251-5G
Methyl 4-(2,2-dicyanovinyl)benzoate
Methyl 4-(2,2-dicyanovinyl)benzoate
Formel:
C₁₂H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01141799 CAS-Nummer: 3129-16-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...