Imidazo[1,2-a]pyridine-6-carbaldehyde oxime
Lieferant: Apollo Scientific
|
Achtung
|
OR111254-1G
APOSOR111254-1GEA
0
EUR
APOSOR111254-1G
Imidazo[1,2-a]pyridine-6-carbaldehyde oxime
Imidazo[1,2-a]pyridine-6-carbaldehyde oxime
Formel:
C₈H₇N₃O Molecular Weight: 161,161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30188177 CAS-Nummer: 1709825-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...