Methyl 2-[4-(3,4-dimethoxyphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]propanoate

Lieferant: Apollo Scientific

OR111256-1G
APOSOR111256-1GEA 0 EUR
APOSOR111256-1G
Methyl 2-[4-(3,4-dimethoxyphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]propanoate
Methyl 2-[4-(3,4-dimethoxyphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]propanoate
Formel: C₁₄H₁₇N₃O₅
Molecular Weight: 307,302 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30481136
CAS-Nummer: 2197054-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR