2-(Chloromethyl)-5-thien-2-yl-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR111257-1G
APOSOR111257-1GEA 0 EUR
APOSOR111257-1G
2-(Chloromethyl)-5-thien-2-yl-1,3,4-oxadiazole
2-(Chloromethyl)-5-thien-2-yl-1,3,4-oxadiazole
Formel: C₇H₅ClN₂OS
Molecular Weight: 200,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01885819
CAS-Nummer: 727374-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR