Methyl 3-chloro-1H-indazole-7-carboxylate

Lieferant: Apollo Scientific
Achtung

OR111267-1G
APOSOR111267-1GEA 331 EUR
APOSOR111267-1G
Methyl 3-chloro-1H-indazole-7-carboxylate
Methyl 3-chloro-1H-indazole-7-carboxylate
Formel: C₉H₇ClN₂O₂
Molecular Weight: 210,617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22376767
CAS-Nummer: 1337881-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR