Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific

OR111270-1G
APOSOR111270-1GEA 0 EUR
APOSOR111270-1G
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6,8-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₀H₈Cl₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02325395
CAS-Nummer: 1431965-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR