3,6-Dichloroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR111273-1G
APOSOR111273-1GEA 371 EUR
APOSOR111273-1G
3,6-Dichloroimidazo[1,2-a]pyridine
3,6-Dichloroimidazo[1,2-a]pyridine
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09994777
CAS-Nummer: 1019027-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR