3,7-Dichloroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR111274-250MG
APOSOR111274-250MGEA 21.9 EUR
APOSOR111274-250MG
3,7-Dichloroimidazo[1,2-a]pyridine
3,7-Dichloroimidazo[1,2-a]pyridine
Formel: C₇H₄Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09994785
CAS-Nummer: 1019026-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR