(1,3-Thiazol-2-ylmethylene)malononitrile

Lieferant: Apollo Scientific

OR111279-1G
APOSOR111279-1GEA 0 EUR
APOSOR111279-1G
(1,3-Thiazol-2-ylmethylene)malononitrile
(1,3-Thiazol-2-ylmethylene)malononitrile
Formel: C₇H₃N₃S
Molecular Weight: 161,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06214304
CAS-Nummer: 771565-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR