4-Methylthiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR111295-250MG
APOSOR111295-250MGEA 174 EUR
APOSOR111295-250MG
4-Methylthiophene-2-carbohydrazide
4-Methylthiophene-2-carbohydrazide
Formel: C₆H₈N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD01627990
CAS-Nummer: 676594-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR