4-Iodo-N-(1-methyl-1H-indazol-3-yl)benzenesulphonamide

Lieferant: Apollo Scientific

OR111297-1G
APOSOR111297-1GEA 339 EUR
APOSOR111297-1G
4-Iodo-N-(1-methyl-1H-indazol-3-yl)benzenesulphonamide
4-Iodo-N-(1-methyl-1H-indazol-3-yl)benzenesulphonamide
Formel: C₁₄H₁₂IN₃O₂S
Molecular Weight: 413,233 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29060037
CAS-Nummer: 2085690-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR