4-Methyl-3,4-dihydro-2H-1,4-benzoxazine
Lieferant: Apollo Scientific
OR111299-1G
APOSOR111299-1GEA
0
EUR
APOSOR111299-1G
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine
Formel:
C₉H₁₁NO Molecular Weight: 149.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01651884 CAS-Nummer: 77901-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...