5'-Brom-2'-hydroxyacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Brom-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-bromphenol

OR1113-1G
APOSOR1113-1GEA 21.9 EUR
APOSOR1113-1G
5'-Brom-2'-hydroxyacetophenon
5'-Brom-2'-hydroxyacetophenon
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 55…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191850
CAS-Nummer: 1450-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR