3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR111302-1G
APOSOR111302-1GEA 232 EUR
APOSOR111302-1G
3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide
3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₈H₆BrClN₄O
Molecular Weight: 289,516 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05974959
CAS-Nummer: 861208-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR