Methyl 2-propyl-1,3-benzoxazole-7-carboxylate

Lieferant: Apollo Scientific

OR111303-1G
APOSOR111303-1GEA 282 EUR
APOSOR111303-1G
Methyl 2-propyl-1,3-benzoxazole-7-carboxylate
Methyl 2-propyl-1,3-benzoxazole-7-carboxylate
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29918598
CAS-Nummer: 2197053-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR