Ethyl imidazo[1,2-a]pyridine-2-carboxylate hydrobromide

Lieferant: Apollo Scientific

OR111305-1G
APOSOR111305-1GEA 0 EUR
APOSOR111305-1G
Ethyl imidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Ethyl imidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Formel: C₁₀H₁₀N₂O₂·BrH
Lagertemperatur: Raumtemperatur
MDL: MFCD12545991
CAS-Nummer: 2549-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR