Ethyl 6-chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR111307-250MG
APOSOR111307-250MGEA 21.9 EUR
APOSOR111307-250MG
Ethyl 6-chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylate
Ethyl 6-chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylate
Formel: C₁₀H₁₀ClN₃O₂
Molecular Weight: 239,658 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11975602
CAS-Nummer: 14714-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR