2,2'-Bis(3-hydroxy-1,4-naphthoquinone)

Lieferant: Apollo Scientific
Achtung

OR11131-1G
APOSOR11131-1GEA 621 EUR
APOSOR11131-1G
2,2'-Bis(3-hydroxy-1,4-naphthoquinone)
2,2'-Bis(3-hydroxy-1,4-naphthoquinone)
Formel: C₂₀H₁₀O₆
Molecular Weight: 346,29 g/mol
Schmelzpunkt: 249…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191788
CAS-Nummer: 33440-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR