Methyl-3-hydroxy-4-iodobenzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-hydroxy-4-iodobenzoate

OR111325-5G
APOSOR111325-5GEA 50.9 EUR
APOSOR111325-5G
Methyl-3-hydroxy-4-iodobenzoate
Methyl-3-hydroxy-4-iodobenzoate
Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203960
CAS-Nummer: 157942-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR