Methyl 3-(benzyloxy)-4-iodobenzoate

Lieferant: Apollo Scientific

OR111328-1G
APOSOR111328-1GEA 0 EUR
APOSOR111328-1G
Methyl 3-(benzyloxy)-4-iodobenzoate
Methyl 3-(benzyloxy)-4-iodobenzoate
Formel: C₁₅H₁₃IO₃
Molecular Weight: 368,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347040
CAS-Nummer: 877064-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR