4-Brom-1,5-dimethyl-1H-pyrazol-3-carbonsäure

Lieferant: Apollo Scientific

OR111332-1G
APOSOR111332-1GEA 0 EUR
APOSOR111332-1G
4-Brom-1,5-dimethyl-1H-pyrazol-3-carbonsäure
4-Bromo-1,5-dimethyl-1H-pyrazole-3-carboxylic acid
Formel: C₆H₇BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02090877
CAS-Nummer: 5775-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR