Ethyl 1-[(3-methoxythien-2-yl)methyl]piperidine-4-carboxylate

Lieferant: Apollo Scientific

OR111355-1G
APOSOR111355-1GEA 0 EUR
APOSOR111355-1G
Ethyl 1-[(3-methoxythien-2-yl)methyl]piperidine-4-carboxylate
Ethyl 1-[(3-methoxythien-2-yl)methyl]piperidine-4-carboxylate
Formel: C₁₄H₂₁NO₃S
Molecular Weight: 283,386 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29918607
CAS-Nummer: 2108826-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR