2-Nitro-1-(3-nitrophenyl)ethanol

Lieferant: Apollo Scientific

OR111360-1G
APOSOR111360-1GEA 0 EUR
APOSOR111360-1G
2-Nitro-1-(3-nitrophenyl)ethanol
2-Nitro-1-(3-nitrophenyl)ethanol
Formel: C₈H₈N₂O₅
Molecular Weight: 212,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25960392
CAS-Nummer: 18731-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR