N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine

Lieferant: Apollo Scientific

OR111366-1G
APOSOR111366-1GEA 0 EUR
APOSOR111366-1G
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
Formel: C₉H₉N₃S
Molecular Weight: 191,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11176831
CAS-Nummer: 63893-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR