(4-Diethylsulphamoyl-phenoxy)-acetic acid

Lieferant: Apollo Scientific

OR111370-1G
APOSOR111370-1GEA 0 EUR
APOSOR111370-1G
(4-Diethylsulphamoyl-phenoxy)-acetic acid
(4-Diethylsulphamoyl-phenoxy)-acetic acid
Formel: C₁₂H₁₇NO₅S
Molecular Weight: 287,332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02708667
CAS-Nummer: 50283-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR