Ethyl 4-bromo-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-bromobenzo[b]thiophene-2-carboxylate

OR111387-1G
APOSOR111387-1GEA 50.9 EUR
APOSOR111387-1G
Ethyl 4-bromo-1-benzothiophene-2-carboxylate
Ethyl 4-bromo-1-benzothiophene-2-carboxylate
Formel: C₁₁H₉BrO₂S
Molecular Weight: 285,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23699513
CAS-Nummer: 93103-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR