4-Bromo-1-benzothiophene-2-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR111388-1G
APOSOR111388-1GEA
387
EUR
APOSOR111388-1G
4-Bromo-1-benzothiophene-2-carbohydrazide
4-Bromo-1-benzothiophene-2-carbohydrazide
Formel:
C₉H₇BrN₂OS Lagertemperatur: Raumtemperatur |
MDL:
MFCD12032264 CAS-Nummer: 1171927-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...