2-(Chloromethyl)-6-methoxy-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

OR111389-100MG
APOSOR111389-100MGEA 91.5 EUR
APOSOR111389-100MG
2-(Chloromethyl)-6-methoxy-1,3-benzoxazole
2-(Chloromethyl)-6-methoxy-1,3-benzoxazole
Formel: C₉H₈ClNO₂
Molecular Weight: 197,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696616
CAS-Nummer: 944897-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR