3-Bromimidazo[1,2-a]pyridin-6-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR111401-250MG
APOSOR111401-250MGEA 21.9 EUR
APOSOR111401-250MG
3-Bromimidazo[1,2-a]pyridin-6-carbonsäure
3-Bromimidazo[1,2-a]pyridin-6-carbonsäure
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241,05 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD07021511
CAS-Nummer: 886362-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR