1-Methyl-1H-indole-3-carboxamide

Lieferant: Apollo Scientific
Achtung

OR111403-1G
APOSOR111403-1GEA 279 EUR
APOSOR111403-1G
1-Methyl-1H-indole-3-carboxamide
1-Methyl-1H-indole-3-carboxamide
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,199 g/mol
Schmelzpunkt: 183…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00457668
CAS-Nummer: 118959-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR