1-Methylindole-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-indole-3-carbonitrile

OR111404-1G
APOSOR111404-1GEA 23.2 EUR
APOSOR111404-1G
1-Methylindole-3-carbonitrile
1-Methylindole-3-carbonitrile
Formel: C₁₀H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00466602
CAS-Nummer: 24662-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR