(2-Bromo-4-methylphenyl)methanol
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-4-methylbenzyl alcohol
OR111405-1G
APOSOR111405-1GEA
0
EUR
APOSOR111405-1G
(2-Bromo-4-methylphenyl)methanol
(2-Bromo-4-methylphenyl)methanol
Formel:
C₈H₉BrO Molecular Weight: 201,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06202838 CAS-Nummer: 824-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...