4-Chloro-2-methylsulphanyl-6-phenylpyrimidine

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-(methylthio)-6-phenylpyrimidine

OR111410-1G
APOSOR111410-1GEA 0 EUR
APOSOR111410-1G
4-Chloro-2-methylsulphanyl-6-phenylpyrimidine
4-Chloro-2-methylsulphanyl-6-phenylpyrimidine
Formel: C₁₁H₉ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD07656565
CAS-Nummer: 91063-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR