4-Hydroxy-2-mercapto-6-thien-2-ylpyrimidine-5-carbonitrile

Lieferant: Apollo Scientific

OR111416-1G
APOSOR111416-1GEA 255 EUR
APOSOR111416-1G
4-Hydroxy-2-mercapto-6-thien-2-ylpyrimidine-5-carbonitrile
4-Hydroxy-2-mercapto-6-thien-2-ylpyrimidine-5-carbonitrile
Formel: C₉H₅N₃OS₂
Molecular Weight: 235,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05843773
CAS-Nummer: 109532-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR