4-Chloro-2-(methylthio)-6-thien-2-ylpyrimidine-5-carbonitrile

Lieferant: Apollo Scientific

OR111418-1G
APOSOR111418-1GEA 0 EUR
APOSOR111418-1G
4-Chloro-2-(methylthio)-6-thien-2-ylpyrimidine-5-carbonitrile
4-Chloro-2-(methylthio)-6-thien-2-ylpyrimidine-5-carbonitrile
Formel: C₁₀H₆ClN₃S₂
Molecular Weight: 267,758 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30718778
CAS-Nummer: 439808-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR