2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Carboxy-1-methyl-1H-pyrrol-2-yl)acetic acid

OR11143-1G
APOSOR11143-1GEA 58 EUR
APOSOR11143-1G
2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid
2-(Carboxymethyl)-1-methyl-1H-pyrrole-3-carboxylic acid
Formel: C₈H₉NO₄
Molecular Weight: 183,161 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00044904
CAS-Nummer: 83863-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR