5-Chlorbenzimidazol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Chloro-1H-benzimidazole
OR11144-1G
APOSOR11144-1GEA
21.9
EUR
APOSOR11144-1G
5-Chlorbenzimidazol
5-Chlorobenzimidazole
Formel:
C₇H₅ClN₂ Molecular Weight: 152.581 g/mol Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD17015012 CAS-Nummer: 4887-82-5 EINECS: 225-503-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...