3-(Dimethylamino)-1-(2-hydroxy-5-methylphenyl)prop-2-en-1-one
Lieferant: Apollo Scientific
OR111449-1G
APOSOR111449-1GEA
0
EUR
APOSOR111449-1G
3-(Dimethylamino)-1-(2-hydroxy-5-methylphenyl)prop-2-en-1-one
3-(Dimethylamino)-1-(2-hydroxy-5-methylphenyl)prop-2-en-1-one
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205,253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10700294 CAS-Nummer: 1292814-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...