5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
Lieferant: Apollo Scientific
OR111451-1G
APOSOR111451-1GEA
0
EUR
APOSOR111451-1G
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
5-Methyl-3-phenyl-4-(1H-pyrazol-5-yl)isoxazole
Formel:
C₁₃H₁₁N₃O Molecular Weight: 225.246 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01765387 CAS-Nummer: 265125-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...