Methyl (4-bromophenyl)(hydroxy)acetate

Lieferant: Apollo Scientific
Achtung

OR111453-1G
APOSOR111453-1GEA 0 EUR
APOSOR111453-1G
Methyl (4-bromophenyl)(hydroxy)acetate
Methyl (4-bromophenyl)(hydroxy)acetate
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16693827
CAS-Nummer: 127709-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR