Ethyl 4-(4-chlorophenyl)-3-methyl-2,4-dioxobutanoate

Lieferant: Apollo Scientific

OR111464-1G
APOSOR111464-1GEA 451 EUR
APOSOR111464-1G
Ethyl 4-(4-chlorophenyl)-3-methyl-2,4-dioxobutanoate
Ethyl 4-(4-chlorophenyl)-3-methyl-2,4-dioxobutanoate
Formel: C₁₃H₁₃ClO₄
Molecular Weight: 268,693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411133
CAS-Nummer: 169544-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR