2-(3,4-Dichlorophenoxy)nicotinonitrile
Lieferant: Apollo Scientific
OR111474-1G
APOSOR111474-1GEA
0
EUR
APOSOR111474-1G
2-(3,4-Dichlorophenoxy)nicotinonitrile
2-(3,4-Dichlorophenoxy)nicotinonitrile
Formel:
C₁₂H₆Cl₂N₂O Molecular Weight: 265,095 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13238892 CAS-Nummer: 99902-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...