7-Methylimidazo[1,2-a]pyridine-8-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR111479-1G
APOSOR111479-1GEA 331 EUR
APOSOR111479-1G
7-Methylimidazo[1,2-a]pyridine-8-carbonitrile
7-Methylimidazo[1,2-a]pyridine-8-carbonitrile
Formel: C₉H₇N₃
Molecular Weight: 157,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22553043
CAS-Nummer: 1352888-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR