2-(Cyclobutylmethoxy)isonicotinonitrile
Lieferant: Apollo Scientific
OR111481-1G
APOSOR111481-1GEA
0
EUR
APOSOR111481-1G
2-(Cyclobutylmethoxy)isonicotinonitrile
2-(Cyclobutylmethoxy)isonicotinonitrile
Formel:
C₁₁H₁₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD13195627 CAS-Nummer: 1235440-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...