2-(Cyclobutylmethoxy)isonicotinic acid
Lieferant: Apollo Scientific
OR111482-1G
APOSOR111482-1GEA
0
EUR
APOSOR111482-1G
2-(Cyclobutylmethoxy)isonicotinic acid
2-(Cyclobutylmethoxy)isonicotinic acid
Formel:
C₁₁H₁₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13195603 CAS-Nummer: 1235440-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...