Ethyl [(3-formyl-1-methyl-1H-indol-2-yl)thio]acetate

Lieferant: Apollo Scientific

OR111483-1G
APOSOR111483-1GEA 0 EUR
APOSOR111483-1G
Ethyl [(3-formyl-1-methyl-1H-indol-2-yl)thio]acetate
Ethyl [(3-formyl-1-methyl-1H-indol-2-yl)thio]acetate
Formel: C₁₄H₁₅NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD31380614
CAS-Nummer: 2197055-31-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR