4-Cyclopropyl-2-pyrimidinamine
Lieferant: Apollo Scientific
|
Achtung
|
OR111493-250MG
APOSOR111493-250MGEA
108
EUR
APOSOR111493-250MG
4-Cyclopropyl-2-pyrimidinamine
4-Cyclopropyl-2-pyrimidinamine
Formel:
C₇H₉N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787899 CAS-Nummer: 21573-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...