3,5-Dimethyl-1-(4-nitrophenyl)-1H-pyrazole

Lieferant: Apollo Scientific

OR111496-10G OR111496-1G OR111496-5G
APOSOR111496-10GEA 0 EUR
APOSOR111496-10G APOSOR111496-1G APOSOR111496-5G
3,5-Dimethyl-1-(4-nitrophenyl)-1H-pyrazole
3,5-Dimethyl-1-(4-nitrophenyl)-1H-pyrazole
Formel: C₁₁H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00187810
CAS-Nummer: 13788-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR