2-(1-Adamantyl)acetohydrazide

Lieferant: Apollo Scientific

OR111497-1G
APOSOR111497-1GEA 125 EUR
APOSOR111497-1G
2-(1-Adamantyl)acetohydrazide
2-(1-Adamantyl)acetohydrazide
Formel: C₁₂H₂₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00185799
CAS-Nummer: 19026-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR